(4-(Benzyloxy)-2-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

296437-1G 296437-500MG 296437-5G
77468-380EA 415.09 USD
77468-380 77481-066 77491-438
(4-(Benzyloxy)-2-methylphenyl)(cyclopropyl)methanol ≥95%
(4-(Benzyloxy)-2-methylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₈H₂₀O₂
Storage Temperature: Ambient
MDL Number: MFCD34889855
CAS Number: 2969326-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR