1-(4-(Benzyloxy)-2-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

296435-1G 296435-500MG 296435-5G
77468-376EA 594.31 USD
77468-376 77481-062 77491-434
1-(4-(Benzyloxy)-2-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-(Benzyloxy)-2-methylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₇H₁₃F₅O₂
MW: 344.2759 g/mol
Storage Temperature: Ambient
CAS Number: 2173102-44-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR