2,3-Dichloro-6-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

29640-1G 29640-500MG 29640-5G
77468-372EA 272.95 USD
77468-372 77481-058 77491-430
2,3-Dichloro-6-methoxybenzoic acid ≥97%
2,3-Dichloro-6-methoxybenzoic acid

Precaución: For research use only

Formula: C₈H₆Cl₂O₃
Storage Temperature: Ambient
MDL Number: MFCD08166717
CAS Number: 55776-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR