4-[2-(3-Bromophenoxy)ethyl]morpholine ≥95%
Supplier: AOBChem USA
|
Warning
|
29632-1G
29632-500MG
29632-5G
77468-368EA
94.76
USD
77468-368
77481-054
77491-426
4-[2-(3-Bromophenoxy)ethyl]morpholine ≥95%
4-[2-(3-Bromophenoxy)ethyl]morpholine
Precaución: For research use only
Formula:
C₁₂H₁₆BrNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD03710161 CAS Number: 435283-95-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...