5-Bromo-1,3-dimethyl-2-(propan-2-yloxy)benzene ≥97%

Supplier: AOBChem USA

Warning

29523-1G 29523-500MG 29523-5G
77468-364EA 138.02 USD
77468-364 77481-050 77491-422
5-Bromo-1,3-dimethyl-2-(propan-2-yloxy)benzene ≥97%
5-Bromo-1,3-dimethyl-2-(propan-2-yloxy)benzene

Precaución: For research use only

Formula: C₁₁H₁₅BrO
MW: 243.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18917821
CAS Number: 95717-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR