(4-butoxy-3,5-dimethylphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

294802-1G 294802-500MG 294802-5G
77468-360EA 415.09 USD
77468-360 77481-046 77491-418
(4-butoxy-3,5-dimethylphenyl)(ethyl)sulfane ≥95%
(4-butoxy-3,5-dimethylphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₁₄H₂₂OS
Storage Temperature: Ambient
CAS Number: 3002425-12-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR