(4-butoxy-3,5-dimethylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

294800-1G 294800-500MG 294800-5G
77468-356EA 415.09 USD
77468-356 77481-042 77491-414
(4-butoxy-3,5-dimethylphenyl)(cyclopropyl)methanol ≥95%
(4-butoxy-3,5-dimethylphenyl)(cyclopropyl)methanol

Precaución: For research use only

Formula: C₁₆H₂₄O₂
MW: 248.3605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889506
CAS Number: 2969436-03-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR