1-(4-butoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

294798-1G 294798-500MG 294798-5G
77468-352EA 415.09 USD
77468-352 77481-038 77491-410
1-(4-butoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-butoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₅H₁₇F₅O₂
MW: 324.2862 g/mol
Storage Temperature: Ambient
CAS Number: 3002522-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR