1-(4-butoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

294796-1G 294796-500MG 294796-5G
77468-348EA 415.09 USD
77468-348 77481-034 77491-406
1-(4-butoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
1-(4-butoxy-3,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₄H₁₈F₂O₂
MW: 256.2883 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889505
CAS Number: 2969419-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR