(4-Ethoxy-3,5-dimethylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

294792-1G 294792-500MG 294792-5G
77468-344EA 415.09 USD
77468-344 77481-030 77491-402
(4-Ethoxy-3,5-dimethylphenyl)(isopropyl)sulfane ≥95%
(4-Ethoxy-3,5-dimethylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₃H₂₀OS
MW: 224.3623 g/mol
Storage Temperature: Ambient
CAS Number: 3002471-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR