(4-Ethoxy-3,5-dimethylphenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

294789-1G 294789-500MG 294789-5G
77468-340EA 415.09 USD
77468-340 77481-026 77491-398
(4-Ethoxy-3,5-dimethylphenyl)trimethylsilane ≥95%
(4-Ethoxy-3,5-dimethylphenyl)trimethylsilane

Precaución: For research use only

Formula: C₁₃H₂₂OSi
MW: 222.3986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889501
CAS Number: 2969435-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR