1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

294785-1G 294785-500MG 294785-5G
77468-336EA 415.09 USD
77468-336 77481-022 77491-394
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-dimethylpropan-1-ol

Precaución: For research use only

Formula: C₁₅H₂₄O₂
MW: 236.3498 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889497
CAS Number: 2969435-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR