Cyclopropyl(4-ethoxy-3,5-dimethylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

294783-1G 294783-500MG 294783-5G
77468-332EA 415.09 USD
77468-332 77481-018 77491-390
Cyclopropyl(4-ethoxy-3,5-dimethylphenyl)methanol ≥95%
Cyclopropyl(4-ethoxy-3,5-dimethylphenyl)methanol

Precaución: For research use only

Formula: C₁₄H₂₀O₂
MW: 220.3074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889495
CAS Number: 2969417-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR