1-(4-Ethoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

294781-1G 294781-500MG 294781-5G
77468-328EA 415.09 USD
77468-328 77481-014 77491-386
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₃H₁₃F₅O₂
MW: 296.2331 g/mol
Storage Temperature: Ambient
CAS Number: 3002523-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR