1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%

Supplier: AOBChem USA

Warning

294779-1G 294779-500MG 294779-5G 294779-25G
77468-324EA 415.09 USD
77468-324 77481-010 77491-382 77501-376
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone ≥95%
1-(4-Ethoxy-3,5-dimethylphenyl)-2,2-difluoroethanone

Precaución: For research use only

Formula: C₁₂H₁₄F₂O₂
MW: 228.2351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34889494
CAS Number: 2969242-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR