1-[2,2'-Bithiophen]-5-ylethanone ≥95%

Supplier: AOBChem USA

Warning

294354-1G 294354-500MG 294354-5G 294354-25G
77468-320EA 86.52 USD
77468-320 77481-006 77491-378 77501-372
1-[2,2'-Bithiophen]-5-ylethanone ≥95%
1-[2,2'-Bithiophen]-5-ylethanone

Precaución: For research use only

Formula: C₁₀H₈OS₂
MW: 208.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00464659
CAS Number: 3515-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR