3-(Ethylthio)-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

294251-1G 294251-500MG 294251-5G
77468-316EA 519.12 USD
77468-316 77481-002 77491-374
3-(Ethylthio)-1,1'-biphenyl ≥95%
3-(Ethylthio)-1,1'-biphenyl

Precaución: For research use only

Formula: C₁₄H₁₄S
MW: 214.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18375504
CAS Number: 1443327-45-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR