1-[2-(2-Propen-1-yl)phenyl]ethanone ≥95%

Supplier: AOBChem USA

Warning

293750-1G 293750-500MG 293750-5G
77468-312EA 434.66 USD
77468-312 77480-998 77491-370
1-[2-(2-Propen-1-yl)phenyl]ethanone ≥95%
1-[2-(2-Propen-1-yl)phenyl]ethanone

Precaución: For research use only

Formula: C₁₁H₁₂O
MW: 160.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24688578
CAS Number: 64664-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR