4-(2-Propen-1-yl)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

293678-1G 293678-500MG 293678-5G
77468-308EA 844.6 USD
77468-308 77480-994 77491-362
4-(2-Propen-1-yl)benzaldehyde ≥95%
4-(2-Propen-1-yl)benzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₀O
MW: 146.19 g/mol
Storage Temperature: Ambient
CAS Number: 77785-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR