5-Chloro-2-isopropylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

29348-1G 29348-500MG 29348-5G
77468-304EA 918.76 USD
77468-304 77480-990 77491-358
5-Chloro-2-isopropylbenzaldehyde ≥97%
5-Chloro-2-isopropylbenzaldehyde

Precaución: For research use only

Formula: C₁₀H₁₁ClO
MW: 182.647 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21705715
CAS Number: 1289090-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR