4-Bromo-3-isopropylphenol ≥95%

Supplier: AOBChem USA

Warning

29328-1G 29328-500MG 29328-5G 29328-25G
77468-300EA 351.23 USD
77468-300 77480-986 77491-354 77501-354
4-Bromo-3-isopropylphenol ≥95%
4-Bromo-3-isopropylphenol

Precaución: For research use only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD15527199
CAS Number: 16606-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR