(4-Ethoxy-2-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

292737-1G 292737-500MG 292737-5G 292737-25G
77468-292EA 415.09 USD
77468-292 77480-978 77491-344 77501-344
(4-Ethoxy-2-methylphenyl)(phenyl)methanol ≥95%
(4-Ethoxy-2-methylphenyl)(phenyl)methanol

Precaución: For research use only

Formula: C₁₆H₁₈O₂
Storage Temperature: Ambient
MDL Number: MFCD34888980
CAS Number: 2969446-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR