(4-Ethoxy-2-methylphenyl)(isopropyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

292731-1G 292731-500MG 292731-5G 292731-25G
77468-288EA 415.09 USD
77468-288 77480-974 77491-340 77501-340
(4-Ethoxy-2-methylphenyl)(isopropyl)sulfane ≥95%
(4-Ethoxy-2-methylphenyl)(isopropyl)sulfane

Precaución: For research use only

Formula: C₁₂H₁₈OS
Storage Temperature: Ambient
MDL Number: MFCD34888976
CAS Number: 2969338-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR