2-Methyl-4-((4-methylbenzyl)oxy)benzoic acid ≥95%

Supplier: AOBChem USA

Warning

292667-1G 292667-500MG 292667-5G 292667-25G
77468-280EA 415.09 USD
77468-280 77480-966 77491-332 77501-332
2-Methyl-4-((4-methylbenzyl)oxy)benzoic acid ≥95%
2-Methyl-4-((4-methylbenzyl)oxy)benzoic acid

Precaución: For research use only

Formula: C₁₆H₁₆O₃
MW: 256.2964 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888928
CAS Number: 1991866-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR