2-Bromo-3-iodo-5-methylpyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-iodo-5-picoline

292612-1G 292612-500MG 292612-5G 292612-25G
77468-276EA 333.72 USD
77468-276 77480-962 77491-326 77501-328
2-Bromo-3-iodo-5-methylpyridine ≥95%
2-Bromo-3-iodo-5-methylpyridine

Precaución: For research use only

Formula: C₆H₅BrIN
MW: 297.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037463
CAS Number: 65550-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR