1-(4-Bromo-3-methylphenyl)piperidine ≥95%

Supplier: AOBChem USA

Warning

29256-1G 29256-500MG 29256-5G 29256-25G
77468-272EA 138.02 USD
77468-272 77480-960 77491-324 77501-326
1-(4-Bromo-3-methylphenyl)piperidine ≥95%
1-(4-Bromo-3-methylphenyl)piperidine

Precaución: For research use only

Formula: C₁₂H₁₆BrN
MW: 254.166 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24133737
CAS Number: 1366131-46-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR