2-(Phenylmethoxy)-4-(trifluoromethoxy)benzaldehyde ≥95%

Supplier: AOBChem USA

Warning

29234-1G 29234-500MG 29234-5G
77468-268EA 541.78 USD
77468-268 77480-956 77491-320
2-(Phenylmethoxy)-4-(trifluoromethoxy)benzaldehyde ≥95%
2-(Phenylmethoxy)-4-(trifluoromethoxy)benzaldehyde

Precaución: For research use only

Formula: C₁₅H₁₁F₃O₃
MW: 296.241 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806028
CAS Number: 1321963-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR