1,4-Dibromo-2,3-dimethoxybenzene ≥95%

Supplier: AOBChem USA

Warning

292162-1G 292162-500MG 292162-5G
77468-264EA 453.2 USD
77468-264 77480-952 77491-316
1,4-Dibromo-2,3-dimethoxybenzene ≥95%
1,4-Dibromo-2,3-dimethoxybenzene

Precaución: For research use only

Formula: C₈H₈Br₂O₂
MW: 295.96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31382357
CAS Number: 175844-47-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR