1-Bromo-2-methoxy-3-(phenylmethoxy)benzene ≥97%

Supplier: AOBChem USA

Warning

29199-1G 29199-500MG 29199-5G 29199-25G
77468-260EA 237.93 USD
77468-260 77480-948 77491-312 77501-322
1-Bromo-2-methoxy-3-(phenylmethoxy)benzene ≥97%
1-Bromo-2-methoxy-3-(phenylmethoxy)benzene

Precaución: For research use only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28806024
CAS Number: 1809161-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR