(3,4-Dichloro-2-fluorophenyl)trimethylsilane ≥95%

Supplier: AOBChem USA

Warning

291391-1G 291391-500MG 291391-5G 291391-25G
77468-256EA 415.09 USD
77468-256 77480-944 77491-308 77501-318
(3,4-Dichloro-2-fluorophenyl)trimethylsilane ≥95%
(3,4-Dichloro-2-fluorophenyl)trimethylsilane

Precaución: For research use only

Formula: C₉H₁₁Cl₂FSi
Storage Temperature: Ambient
CAS Number: 3002532-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR