Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

291387-1G 291387-500MG 291387-5G 291387-25G
77468-252EA 415.09 USD
77468-252 77480-940 77491-304 77501-314
Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanol ≥95%
Cyclopropyl(3,4-dichloro-2-fluorophenyl)methanol

Precaución: For research use only

Formula: C₁₀H₉Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD34888115
CAS Number: 2969385-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR