(3,4-Dichloro-2-fluorophenyl)triethylsilane ≥95%

Supplier: AOBChem USA

Warning

291385-1G 291385-500MG 291385-5G 291385-25G
77468-248EA 415.09 USD
77468-248 77480-936 77491-300 77501-310
(3,4-Dichloro-2-fluorophenyl)triethylsilane ≥95%
(3,4-Dichloro-2-fluorophenyl)triethylsilane

Precaución: For research use only

Formula: C₁₂H₁₇Cl₂FSi
MW: 279.2532 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888113
CAS Number: 2969342-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR