1-(3,4-Dichloro-2-fluorophenyl)cyclobutanol ≥95%

Supplier: AOBChem USA

Warning

291383-1G 291383-500MG 291383-5G 291383-25G
77468-244EA 415.09 USD
77468-244 77480-932 77491-296 77501-306
1-(3,4-Dichloro-2-fluorophenyl)cyclobutanol ≥95%
1-(3,4-Dichloro-2-fluorophenyl)cyclobutanol

Precaución: For research use only

Formula: C₁₀H₉Cl₂FO
Storage Temperature: Ambient
MDL Number: MFCD34888112
CAS Number: 2969444-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR