1-(3,4-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

291381-1G 291381-500MG 291381-5G 291381-25G
77468-240EA 415.09 USD
77468-240 77480-928 77491-292 77501-302
1-(3,4-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(3,4-Dichloro-2-fluorophenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₈H₂Cl₂F₄O
Storage Temperature: Ambient
MDL Number: MFCD34888110
CAS Number: 2969474-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR