1-(4-Chloro-2,3-difluorophenyl)-2-fluoroethanone ≥95%

Supplier: AOBChem USA

Warning

291373-1G 291373-500MG 291373-5G 291373-25G
77468-236EA 415.09 USD
77468-236 77480-924 77491-288 77501-298
1-(4-Chloro-2,3-difluorophenyl)-2-fluoroethanone ≥95%
1-(4-Chloro-2,3-difluorophenyl)-2-fluoroethanone

Precaución: For research use only

Formula: C₈H₄ClF₃O
MW: 208.5649 g/mol
Storage Temperature: Ambient
CAS Number: 3002523-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR