1-(4-Chloro-2,3-difluorophenyl)-2,2,2-trifluoroethanone ≥95%

Supplier: AOBChem USA

Warning

291369-1G 291369-500MG 291369-5G 291369-25G
77468-232EA 415.09 USD
77468-232 77480-920 77491-284 77501-294
1-(4-Chloro-2,3-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-Chloro-2,3-difluorophenyl)-2,2,2-trifluoroethanone

Precaución: For research use only

Formula: C₈H₂ClF₅O
MW: 244.5458 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888101
CAS Number: 2969431-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR