1-(2,3-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%

Supplier: AOBChem USA

Warning

291364-1G 291364-500MG 291364-5G 291364-25G
77468-228EA 415.09 USD
77468-228 77480-914 77491-278 77501-290
1-(2,3-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one ≥95%
1-(2,3-Difluoro-4-methoxyphenyl)-2,2,3,3,3-pentafluoropropan-1-one

Precaución: For research use only

Formula: C₁₀H₅F₇O₂
MW: 290.1343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34888097
CAS Number: 2969444-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR