(2,3-Difluoro-4-methoxyphenyl)(ethyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

291362-1G 291362-500MG 291362-5G 291362-25G
77468-224EA 415.09 USD
77468-224 77480-910 77491-274 77501-284
(2,3-Difluoro-4-methoxyphenyl)(ethyl)sulfane ≥95%
(2,3-Difluoro-4-methoxyphenyl)(ethyl)sulfane

Precaución: For research use only

Formula: C₉H₁₀F₂OS
Storage Temperature: Ambient
CAS Number: 3002532-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR