4-Bromo-2-nitro-1-phenoxybenzene ≥95%

Supplier: AOBChem USA

Warning

29054-1G 29054-500MG 29054-5G 29054-25G
77468-220EA 134.93 USD
77468-220 77480-908 77491-272 77501-282
4-Bromo-2-nitro-1-phenoxybenzene ≥95%
4-Bromo-2-nitro-1-phenoxybenzene

Precaución: For research use only

Formula: C₁₂H₈BrNO₃
Storage Temperature: Ambient
MDL Number: MFCD13191935
CAS Number: 56966-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR