5-(Hydroxymethyl)-2-phenoxypyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (6-Phenoxy-3-pyridinyl)methanol, (6-Phenoxypyridin-3-yl)methanol

29026-1G 29026-500MG 29026-5G 29026-25G
77468-216EA 138.02 USD
77468-216 77480-904 77491-268 77501-278
5-(Hydroxymethyl)-2-phenoxypyridine ≥95%
5-(Hydroxymethyl)-2-phenoxypyridine

Precaución: For research use only

Formula: C₁₂H₁₁NO₂
MW: 201.221 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02682069
CAS Number: 101990-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR