[1,1':3',1''-terphenyl]-5'-yl(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

290035-1G 290035-500MG 290035-5G 290035-25G
77468-208EA 415.09 USD
77468-208 77480-898 77491-260 77501-270
[1,1':3',1''-terphenyl]-5'-yl(methyl)sulfane ≥95%
[1,1':3',1''-terphenyl]-5'-yl(methyl)sulfane

Precaución: For research use only

Formula: C₁₉H₁₆S
Storage Temperature: Ambient
MDL Number: MFCD34887117
CAS Number: 2969324-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR