1-Methoxy-2-(methoxymethoxy)benzene ≥95%

Supplier: AOBChem USA

Warning

28883-1G 28883-5G 28883-25G
77468-204EA 60.77 USD
77468-204 77491-254 77501-264
1-Methoxy-2-(methoxymethoxy)benzene ≥95%
1-Methoxy-2-(methoxymethoxy)benzene

Precaución: For research use only

Formula: C₉H₁₂O₃
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27928051
CAS Number: 73220-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR