5-Bromo-2-(4-methylphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

28864-1G 28864-500MG 28864-5G 28864-25G
77468-200EA 368.74 USD
77468-200 77480-892 77491-250 77501-260
5-Bromo-2-(4-methylphenyl)pyridine ≥97%
5-Bromo-2-(4-methylphenyl)pyridine

Precaución: For research use only

Formula: C₁₂H₁₀BrN
MW: 248.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14707252
CAS Number: 867380-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR