4-(1,3-Dioxolan-2-yl)benzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
28782-1G
28782-500MG
28782-5G
28782-25G
77468-196EA
197.76
USD
77468-196
77480-888
77491-244
77501-250
4-(1,3-Dioxolan-2-yl)benzaldehyde ≥97%
4-(1,3-Dioxolan-2-yl)benzaldehyde
Precaución: For research use only
Formula:
C₁₀H₁₀O₃ Storage Temperature: Ambient |
MDL Number:
MFCD03180272 CAS Number: 40681-88-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...