2-(4-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane ≥95%

Supplier: AOBChem USA

Warning

28760-1G 28760-500MG 28760-5G 28760-25G
77468-192EA 422.3 USD
77468-192 77480-884 77491-240 77501-246
2-(4-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane ≥95%
2-(4-Bromo-2-fluoro-6-methoxyphenyl)-1,3-dioxolane

Precaución: For research use only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619423
CAS Number: 2221812-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR