Cyclopropyl(4'-methyl[1,1'-biphenyl]-4-yl)methanone ≥95%

Supplier: AOBChem USA

Warning

287161-1G 287161-500MG 287161-5G 287161-25G
77468-188EA 415.09 USD
77468-188 77480-880 77491-236 77501-244
Cyclopropyl(4'-methyl[1,1'-biphenyl]-4-yl)methanone ≥95%
Cyclopropyl(4'-methyl[1,1'-biphenyl]-4-yl)methanone

Precaución: For research use only

Formula: C₁₇H₁₆O
Storage Temperature: Ambient
MDL Number: MFCD11918943
CAS Number: 203066-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR