Cyclopropyl[4-(ethylthio)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287118-1G 287118-500MG 287118-5G
77468-184EA 576.8 USD
77468-184 77480-876 77491-232
Cyclopropyl[4-(ethylthio)phenyl]methanone ≥95%
Cyclopropyl[4-(ethylthio)phenyl]methanone

Precaución: For research use only

Formula: C₁₂H₁₄OS
MW: 206.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19251805
CAS Number: 1267833-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR