Cyclopropyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287115-1G 287115-500MG 287115-5G
77468-180EA 546.93 USD
77468-180 77480-872 77491-228
Cyclopropyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[2-fluoro-5-(trifluoromethyl)phenyl]methanone

Precaución: For research use only

Formula: C₁₁H₈F₄O
MW: 232.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23823061
CAS Number: 1506501-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR