Cyclopropyl[2-fluoro-3-(trifluoromethyl)phenyl]methanone ≥95%

Supplier: AOBChem USA

Warning

287086-1G 287086-500MG 287086-5G
77468-176EA 567.53 USD
77468-176 77480-868 77491-224
Cyclopropyl[2-fluoro-3-(trifluoromethyl)phenyl]methanone ≥95%
Cyclopropyl[2-fluoro-3-(trifluoromethyl)phenyl]methanone

Precaución: For research use only

Formula: C₁₁H₈F₄O
Storage Temperature: Ambient
MDL Number: MFCD23823173
CAS Number: 1502713-38-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR