(4-Butylphenyl)cyclopropylmethanone ≥95%

Supplier: AOBChem USA

Warning

287069-1G 287069-500MG 287069-5G
77468-172EA 457.32 USD
77468-172 77480-864 77491-220
(4-Butylphenyl)cyclopropylmethanone ≥95%
(4-Butylphenyl)cyclopropylmethanone

Precaución: For research use only

Formula: C₁₄H₁₈O
MW: 202.29 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11210435
CAS Number: 1094416-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR